Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145600.1 | NP_663575.1 | 6, 7 | 511 | ||
GenBank mRNA | AK029489.1 | - | 5, 6 | 336 | ||
AK037748.1 | - | 2, 3 | 793 | |||
AK050534.1 | - | 6, 7 | 521 | |||
AK086617.1 | - | 6, 7 | 545 | |||
BC008086.1 | - | 6, 7 | 511 | 3586161 | ||
BC030675.1 | - | 6, 7 | 509 | 1068780 | ||
BC037025.1 | - | 2, 3 | 1868 | |||
BC083081.1 | - | 6, 7 | 545 | 6510396 |