Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195006.1 | NP_001181935.1 | 15 | 1452 | ||
NM_145602.3 | NP_663577.1 | 14 | 1413 | |||
GenBank mRNA | AK090374.1 | - | 15 | 1434 | ||
AK144294.1 | - | 15 | 1435 | |||
AK168873.1 | - | 15 | 1436 | |||
BC006595.1 | - | 14 | 1387 | 3156802 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK129305.1 | - | 1747 | Chr. - - [-] on Build |