Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D230025D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145604.2 | NP_663579.2 | 3, 4 | 474 | ||
GenBank mRNA | AK051965.1 | - | 3, 4 | 460 | ||
AK156120.1 | - | 3, 4 | 476 | |||
AK166937.1 | - | 3, 4 | 457 | |||
AK213296.1 | - | 1, 2 | 33 | |||
AK218692.1 | - | 2, 3 | 117 | |||
BC006874.1 | - | 3, 4 | 357 | 2646515 | ||
BC057919.1 | - | 3, 4 | 463 | 5134302 | ||
BC066141.1 | - | 3, 4 | 395 | |||
BC066826.1 | - | 3, 4 | 395 | |||
CN703950.1 | - | 3, 4 | 474 |