Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145619.2 | NP_663594.2 | 4, 5 | 654 | ||
GenBank mRNA | AF368233.1 | - | 4, 5 | 654 | ||
AK147868.1 | - | 4, 5 | 953 | |||
AK170541.1 | - | 4, 5 | 598 | |||
AK182093.1 | - | 80 |