Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167775.1 | NP_001161247.1 | 3 | 865 | ||
NM_001167776.1 | NP_001161248.1 | 3 | 578 | |||
NM_145627.2 | NP_663602.1 | 3 | 578 | |||
GenBank mRNA | AK041871.1 | - | 3 | 553 | ||
AK089105.1 | - | 3 | 389 | |||
AK122208.1 | - | 3 | 496 | |||
AK145991.1 | - | 3 | 578 | |||
AK153736.1 | - | 3 | 866 | |||
AK167666.1 | - | 3 | 556 | |||
AK186699.1 | - | 38 | ||||
AK187170.1 | - | 38 | ||||
BC004674.1 | - | 2 | 263 | 3500403 | ||
BC093519.1 | - | 3 | 510 |
Gene Symbol: Gm15455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161816.1 | NP_001155288.1 | 1 | 395 | ||
GenBank mRNA | AK139531.1 | - | 1144 | |||
BC157984.1 | - | 1 | 319 | |||
BC157993.1 | - | 1 | 319 |