Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300lf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169153.1 | NP_001162624.1 | 6 | 923 | ||
NM_145634.3 | NP_663609.2 | 6 | 902 | |||
GenBank mRNA | AB292061.1 | - | 6 | 812 | ||
AF251703.1 | - | 5 | 715 | |||
AK089313.1 | - | 6 | 923 | |||
AK154273.1 | - | 5 | 733 | |||
AK155367.1 | - | 6 | 900 | |||
AY214460.1 | - | 6 | 854 | |||
AY457047.1 | - | 6 | 812 | |||
BC057864.1 | - | 6 | 867 | 5318856 | ||
JX073136.1 | - | 5 | 768 |