Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145706.2 | NP_663752.2 | 7, 8 | 982 | ||
GenBank mRNA | AK012191.1 | - | 7, 8 | 976 | ||
AK090019.1 | - | 7, 8 | 945 | |||
AK146262.1 | - | 7, 8 | 984 | |||
BC011060.1 | - | 7, 8 | 970 | 3156436 | ||
BC052530.1 | - | 7, 8 | 910 | |||
DV046453.1 | - | 455 |