Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tardbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003898.3 | NP_001003898.1 | 6 | 1076 | ||
NM_001003899.2 | NP_001003899.1 | 6 | 1076 | |||
NM_001008545.2 | NP_001008545.1 | 6 | 1076 | |||
NM_001008546.2 | NP_001008546.1 | 6 | 1076 | |||
NM_145556.4 | NP_663531.1 | 6 | 1076 | |||
GenBank mRNA | AK032199.1 | - | 6 | 846 | ||
AK045495.1 | - | 6 | 846 | |||
AK076208.1 | - | 6 | 844 | |||
AK083312.1 | - | 6 | 850 | |||
AK133207.1 | - | 6 | 853 | |||
AK146002.1 | - | 6 | 846 | |||
AK146997.1 | - | 7 | 755 | |||
AK153803.1 | - | 6 | 850 | |||
AK194463.1 | - | 2 | 152 | |||
AK195880.1 | - | 2 | 158 | |||
AY319513.1 | - | 5 | 746 | |||
AY319514.1 | - | 5 | 746 | |||
AY319515.1 | - | 5 | 746 | |||
AY319516.1 | - | 5 | 746 | |||
BC012873.1 | - | 6 | 834 | 4016437 | ||
BC025544.1 | - | 6 | 847 | 5321007 | ||
BC027105.1 | - | 6 | 838 | 5359082 | ||
BC027772.1 | - | 6 | 836 | 5346061 | ||
BC029728.1 | - | 6 | 837 | 4950263 | ||
BC031126.1 | - | 6 | 838 | 4977114 | ||
BC033475.1 | - | 6 | 834 | 4953334 | ||
GQ428775.1 | - | 5 | 746 | |||
GQ428776.1 | - | 5 | 746 | |||
GQ428777.1 | - | 5 | 746 | |||
GU250735.1 | - | 5 | 746 |