Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwh43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181323.2 | NP_851840.1 | 16 | 2333 | ||
GenBank mRNA | AK035550.1 | - | 15 | 1926 | ||
AK081976.1 | - | 16 | 2333 | |||
BC016523.1 | - | 16 | 2161 | 3584006 |