Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145585.2 | NP_663560.1 | 3 | 645 | ||
GenBank mRNA | AK032078.1 | - | 3 | 621 | ||
AK152912.1 | - | 3 | 618 | |||
AK178112.1 | - | 1 | 71 | |||
AK183558.1 | - | 2 | 365 | |||
BC004776.1 | - | 3 | 612 | 3257051 |