Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cngb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145601.1 | NP_663576.1 | 13 | 1117 | ||
GenBank mRNA | AK080705.1 | - | 11 | 2171 | ||
BC016201.1 | - | 13 | 1117 | 4527347 | ||
BC051415.1 | - | 11 | 2026 | |||
BC058272.1 | - | 11 | 1716 |