Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D230025D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145604.2 | NP_663579.2 | 16 | 1634 | ||
GenBank mRNA | AK051965.1 | - | 15 | 1522 | ||
AK156120.1 | - | 16 | 1636 | |||
AK181184.1 | - | 1 | 232 | |||
AK194632.1 | - | 1 | 232 | |||
AK196070.1 | - | 1 | 235 | |||
AK212320.1 | - | 1 | 238 | |||
BC006874.1 | - | 16 | 1517 | 2646515 | ||
BC057919.1 | - | 16 | 1623 | 5134302 | ||
BC066141.1 | - | 11 | 1125 | |||
BC066826.1 | - | 11 | 1125 |