Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145612.2 | NP_663587.1 | 8 | 1782 | ||
GenBank mRNA | AK032556.1 | - | 8 | 1822 | ||
BC005471.1 | - | 8 | 1782 | 3496918 |