Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145619.2 | NP_663594.2 | 11 | 1911 | ||
GenBank mRNA | AF368233.1 | - | 11 | 1911 | ||
AK044223.1 | - | 11 | 1904 | |||
AK088125.1 | - | 11 | 1917 | |||
AK145259.1 | - | 11 | 1945 | |||
AK147868.1 | - | 11 | 2210 | |||
AK165070.1 | - | 9 | 2163 | |||
AK170541.1 | - | 11 | 1855 | |||
AK181166.1 | - | 1 | 267 | |||
BC014870.1 | - | 11 | 1851 | 3602116 | ||
BC058754.1 | - | 11 | 1831 | 6476689 | ||
BC062204.1 | - | 11 | 1830 |