Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm15455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161816.1 | NP_001155288.1 | 1 | 1910 | ||
GenBank mRNA | BC157984.1 | - | 1 | 1834 | ||
BC157993.1 | - | 1 | 1834 |
Gene Symbol: Rbm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167775.1 | NP_001161247.1 | 17 | 2611 | ||
NM_001167776.1 | NP_001161248.1 | 16 | 2096 | |||
NM_145627.2 | NP_663602.1 | 17 | 2327 | |||
GenBank mRNA | AK089105.1 | - | 16 | 1907 | ||
AK122208.1 | - | 14 | 4027 | |||
AK145991.1 | - | 17 | 2327 | |||
AK153736.1 | - | 17 | 2612 | |||
AK167666.1 | - | 16 | 2074 | |||
BC004674.1 | - | 16 | 2012 | 3500403 | ||
BC093519.1 | - | 16 | 2025 |