Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145628.4 | NP_663603.3 | 13 | 1657 | ||
GenBank mRNA | AK031767.1 | - | 13 | 1636 | ||
AK049053.1 | - | 13 | 2071 | |||
AK169116.1 | - | 5 | 480 | |||
AK220478.1 | - | 12 | 1370 | |||
BC005470.1 | - | 11 | 1226 | 3496607 |