Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox12e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145684.1 | NP_663717.1 | 4 | 580 | ||
GenBank mRNA | AK138743.1 | - | 4 | 534 | ||
BC013751.1 | - | 4 | 560 | 3596809 | ||
BC051047.1 | - | 4 | 564 | 4457881 | ||
U39200.1 | - | 4 | 521 | |||
X99252.1 | - | 4 | 580 |