Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17re | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034029.1 | NP_001029201.1 | 2 | 174 | ||
NM_001034031.1 | NP_001029203.1 | 4 | 249 | |||
NM_145826.5 | NP_665825.2 | 3 | 601 | |||
GenBank mRNA | AF458068.1 | - | 3 | 251 | ||
AK046302.1 | - | 3 | 604 | |||
AK046713.1 | - | 3 | 263 | |||
AK169498.1 | - | 3 | 429 | |||
BC069861.1 | - | 3 | 278 | 6311568 | ||
DQ092339.1 | - | 3 | 190 | |||
DQ092340.1 | - | 2 | 174 | |||
DQ092341.1 | - | 4 | 249 |