Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145838.1 | NP_665837.1 | 2, 3 | 278 | ||
GenBank mRNA | AB059554.1 | - | 2, 3 | 278 | ||
AK085105.1 | - | 3, 4 | 415 | |||
AK156557.1 | - | 2, 3 | 815 | |||
AK172400.1 | - | 2, 3 | 343 | |||
BC115912.1 | - | 2, 3 | 233 | 40104779 | ||
BC118063.1 | - | 2, 3 | 233 | 40104780 |