Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspear | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287074.1 | NP_001274003.1 | 3 | 754 | ||
GenBank mRNA | AJ487520.1 | - | 2 | 425 | ||
AK082868.1 | - | 1 | 226 | |||
JQ815565.1 | - | 3 | 754 |