Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145857.2 | NP_665856.2 | 2 | 151 | ||
GenBank mRNA | AF520774.1 | - | 2 | 196 | ||
AK089390.1 | - | 2 | 115 | |||
AY160220.1 | - | 1 | 80 | |||
AY160221.1 | - | 2 | 152 | |||
BC044774.2 | - | 2 | 112 |