Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145916.2 | NP_666021.2 | 3, 4 | 260 | ||
GenBank mRNA | AK143414.1 | - | 2, 3 | 1600 | ||
AK144887.1 | - | 3, 4 | 472 | |||
AK168948.1 | - | 3, 4 | 215 | |||
BC011501.1 | - | 3, 4 | 278 | 3584546 | ||
BC062103.1 | - | 3, 4 | 202 | |||
U29499.1 | - | 1, 2 | 57 |