Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110271.1 | NP_001103741.1 | 3 | 630 | ||
NM_001110272.1 | NP_001103742.1 | 2 | 321 | |||
NM_145919.2 | NP_666031.2 | 4 | 506 | |||
GenBank mRNA | AB073619.1 | - | 4 | 454 | ||
AK003653.1 | - | 3 | 516 | |||
BC006879.1 | - | 4 | 463 | 2647263 | ||
BY144149.1 | - | 2 | 322 | |||
CB952955.1 | - | 2 | 224 |