Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252505.1 | NP_001239434.1 | 4 | 613 | ||
NM_001252506.1 | NP_001239435.1 | 3 | 480 | |||
NM_145933.4 | NP_666045.1 | 4 | 944 | |||
GenBank mRNA | AK084124.1 | - | 4 | 779 | ||
AK196570.1 | - | 1 | 145 | |||
BC027833.1 | - | 1 | 141 | 5321763 | ||
BC092222.1 | - | 4 | 922 | 6844069 | ||
BC096026.1 | - | 4 | 592 | 6833540 | ||
CD349601.1 | - | 4 | 582 | |||
CN458528.1 | - | 1 | 160 | |||
D16106.1 | - | 3 | 431 |