Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145939.2 | NP_666051.2 | 1, 2 | 207 | ||
GenBank mRNA | BC031893.1 | - | 1, 2 | 202 | 5371807 | |
CA538864.1 | - | 1, 2 | 207 | |||
CX217502.1 | - | 1, 2 | 154 |