Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ido2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145949.2 | NP_666061.3 | 3, 4 | 408 | ||
GenBank mRNA | BC026393.1 | - | 3, 4 | 408 | 4195134 | |
EF137182.1 | - | 3, 4 | 408 | |||
EU440733.1 | - | 2, 3 | 200 |