Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145950.4 | NP_666062.3 | 6 | 912 | ||
GenBank mRNA | AK048731.1 | - | 6 | 913 | ||
AK148243.1 | - | 6 | 887 | |||
AK169678.1 | - | 6 | 857 | |||
AK177160.1 | - | 1 | 88 | |||
BC010311.1 | - | 2 | 337 | 3595088 |