Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC027231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145972.4 | NP_666084.1 | 2, 3 | 331 | ||
GenBank mRNA | AK137274.1 | - | 1, 2 | 1024 | ||
AK144612.1 | - | 2, 3 | 312 | |||
AK162291.1 | - | 1, 2 | 173 | |||
BB651637.1 | - | 2, 3 | 329 | |||
BC027231.1 | - | 2, 3 | 334 | 3585175 |