Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000821.6 | NP_000812.2 | 6 | 790 | ||
NM_001142269.3 | NP_001135741.1 | 5 | 619 | |||
XM_005264259.4 | XP_005264316.1 | 6 | 749 | |||
XM_011532764.2 | XP_011531066.1 | 6 | 749 | |||
XM_011532765.2 | XP_011531067.1 | 5 | 588 | |||
XM_017003803.1 | XP_016859292.1 | 5 | 612 | |||
GenBank mRNA | AK222827.1 | - | 6 | 750 | ||
AK225663.1 | - | 6 | 749 | |||
AK297397.1 | - | 5 | 559 | |||
AK298982.1 | - | 5 | 513 | |||
AK310483.1 | - | 6 | 658 | |||
AK313126.1 | - | 6 | 758 | |||
BC013979.2 | - | 6 | 712 | 4129593 | ||
EU847509.1 | - | 6 | 687 | |||
L17128.1 | - | 6 | 839 | |||
M81592.1 | - | 6 | 770 |