Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286573.1 | NP_001273502.1 | 5 | 847 | ||
NM_001286575.1 | NP_001273504.1 | 6 | 683 | |||
NM_145830.2 | NP_665829.1 | 5 | 847 | |||
NM_147151.2 | NP_671493.1 | 6 | 683 | |||
GenBank mRNA | AB077209.1 | - | 6 | 676 | ||
AB077210.1 | - | 5 | 847 | |||
AK154246.1 | - | 5 | 870 | |||
BC058357.1 | - | 6 | 681 |