Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145841.2 | NP_665840.2 | 2 | 969 | ||
GenBank mRNA | AK041231.1 | - | 2 | 382 | ||
AK048960.1 | - | 2 | 933 | |||
AK136779.1 | - | 2 | 970 | |||
AY095374.1 | - | 1 | 185 | |||
AY250713.1 | - | 1 | 185 | |||
BC104048.1 | - | 1 | 185 | 40040143 | ||
BC104049.2 | - | 1 | 185 | 40040144 |