Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pttg1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145925.2 | NP_666037.1 | 3 | 246 | ||
GenBank mRNA | AK035696.1 | - | 3 | 261 | ||
AK075773.1 | - | 3 | 219 | |||
AK088813.1 | - | 3 | 283 | |||
AK089887.1 | - | 3 | 228 | |||
AK098093.1 | - | 3 | 260 | |||
AK159987.1 | - | 3 | 228 | |||
AK167972.1 | - | 3 | 258 | |||
AK168164.1 | - | 3 | 258 | |||
AK170802.1 | - | 3 | 228 | |||
AK170971.1 | - | 3 | 261 | |||
AK170972.1 | - | 3 | 228 | |||
AK171140.1 | - | 3 | 227 | |||
AK199936.1 | - | 3 | 108 | |||
BC023961.1 | - | 3 | 301 | 5323397 | ||
BC025533.1 | - | 3 | 270 | 5250502 | ||
BC029144.1 | - | 3 | 242 | 4945974 | ||
BC032184.1 | - | 3 | 265 | 5310191 |