Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145929.2 | NP_666041.1 | 5 | 723 | ||
GenBank mRNA | AK080881.1 | - | 5 | 346 | ||
AK155195.1 | - | 5 | 363 | |||
AK159771.1 | - | 5 | 346 | |||
AK170437.1 | - | 5 | 363 | |||
AK182264.1 | - | 1 | 24 | |||
AK213106.1 | - | 1 | 23 | |||
BC026802.1 | - | 5 | 723 | 4223300 | ||
BC054422.1 | - | 5 | 367 |