Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145940.2 | NP_666052.1 | 3 | 356 | ||
GenBank mRNA | AK031672.1 | - | 3 | 365 | ||
AK034907.1 | - | 3 | 400 | |||
AK045131.1 | - | 3 | 365 | |||
AK050495.1 | - | 3 | 364 | |||
AK159608.1 | - | 3 | 385 | |||
AK170563.1 | - | 3 | 400 | |||
AK186984.1 | - | 1 | 28 | |||
AK203625.1 | - | 1 | 28 | |||
BC024811.1 | - | 3 | 324 | |||
BC024883.1 | - | 3 | 320 | |||
BC025560.1 | - | 3 | 356 | 5322999 |