Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134465.2 | NP_001127937.1 | 1, 2 | 321 | ||
NM_145969.4 | NP_666081.1 | 1, 2 | 321 | |||
GenBank mRNA | AK054197.1 | - | 1, 2 | 265 | ||
AK054295.1 | - | 1, 2 | 322 | |||
AK131163.1 | - | 1, 2 | 293 | |||
AK132219.1 | - | 1, 2 | 318 | |||
AK161908.1 | - | 1, 2 | 292 | |||
BB666315.1 | - | 1, 2 | 322 | |||
BC025603.1 | - | 1, 2 | 316 | 5250476 |