Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145825.2 | NP_665824.1 | 5 | 560 | ||
GenBank mRNA | AF362367.1 | - | 5 | 560 | ||
AK078275.1 | - | 4 | 2475 | |||
AK090064.1 | - | 418 | ||||
AK131808.1 | - | 5 | 549 | |||
AK166515.1 | - | 3 | 338 | |||
BC038154.1 | - | 4 | 451 | |||
BC060991.1 | - | 5 | 512 | 6742836 | ||
BC087905.1 | - | 5 | 538 | 6707456 |