Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000879.2 | NP_000870.1 | 4 | 493 | ||
XM_005271988.3 | XP_005272045.1 | 5 | 627 | |||
XM_011543373.2 | XP_011541675.1 | 6 | 729 | |||
XM_011543374.2 | XP_011541676.1 | 5 | 680 | |||
GenBank mRNA | X04688.1 | - | 4 | 493 | ||
X12705.1 | - | 4 | 493 |