Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110271.1 | NP_001103741.1 | 6 | 1328 | ||
NM_001110272.1 | NP_001103742.1 | 5 | 1073 | |||
NM_145919.2 | NP_666031.2 | 7 | 1204 | |||
GenBank mRNA | AB073619.1 | - | 7 | 1152 | ||
AK003653.1 | - | 6 | 1213 | |||
BC006879.1 | - | 7 | 1161 | 2647263 | ||
CB055425.1 | - | 2 | 512 |