Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pttg1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145925.2 | NP_666037.1 | 6 | 1835 | ||
GenBank mRNA | AK035696.1 | - | 6 | 1850 | ||
AK075773.1 | - | 6 | 1808 | |||
AK081798.1 | - | 1 | 1258 | |||
AK088813.1 | - | 6 | 1873 | |||
AK089887.1 | - | 6 | 1817 | |||
AK098093.1 | - | 6 | 1849 | |||
AK151639.1 | - | 6 | 1818 | |||
AK152366.1 | - | 6 | 1817 | |||
AK159987.1 | - | 6 | 1817 | |||
AK167972.1 | - | 6 | 1846 | |||
AK168164.1 | - | 6 | 1846 | |||
AK170802.1 | - | 6 | 1817 | |||
AK170971.1 | - | 6 | 1850 | |||
AK170972.1 | - | 6 | 1817 | |||
AK171140.1 | - | 6 | 1816 | |||
AK189329.1 | - | 1 | 31 | |||
AK191037.1 | - | 1 | 255 | |||
AK192116.1 | - | 1 | 255 | |||
AK202715.1 | - | 1 | 255 | |||
AK209759.1 | - | 1 | 255 | |||
AK211153.1 | - | 1 | 256 | |||
AK216317.1 | - | 1 | 255 | |||
AK216585.1 | - | 1 | 375 | |||
BC023961.1 | - | 6 | 1890 | 5323397 | ||
BC025533.1 | - | 6 | 1859 | 5250502 | ||
BC029144.1 | - | 6 | 1831 | 4945974 | ||
BC032184.1 | - | 6 | 1854 | 5310191 |