Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145947.2 | NP_666059.2 | 11 | 1392 | ||
GenBank mRNA | AF345194.1 | - | 10 | 1294 | ||
AK054327.1 | - | 11 | 1393 | |||
BC026928.1 | - | 11 | 1371 | 4020760 |