Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ido2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145949.2 | NP_666061.3 | 10 | 2274 | ||
GenBank mRNA | AK082385.1 | - | 9 | 2465 | ||
BC026393.1 | - | 10 | 2272 | 4195134 | ||
EF137182.1 | - | 10 | 2272 |