Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271447.1 | NP_001258376.1 | 7 | 994 | ||
NM_001271448.1 | NP_001258377.1 | 6 | 839 | |||
NM_001271449.1 | NP_001258378.1 | 7 | 983 | |||
NM_001271450.1 | NP_001258379.1 | 7 | 983 | |||
NM_001271451.1 | NP_001258380.1 | 7 | 983 | |||
NM_145951.5 | NP_666063.1 | 7 | 938 | |||
GenBank mRNA | AK028343.1 | - | 7 | 984 | ||
AK034058.1 | - | 7 | 939 | |||
AK041822.1 | - | 7 | 984 | |||
AK133956.1 | - | 6 | 810 | |||
AK137736.1 | - | 7 | 965 | |||
AK139897.1 | - | 7 | 965 | |||
BC025915.1 | - | 7 | 957 | 5043650 |