Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145969.4 | NP_666081.1 | 18 | 1704 | ||
GenBank mRNA | AK161908.1 | - | 18 | 1674 | ||
BC025603.1 | - | 18 | 1699 | 5250476 |