Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282055.1 | NP_001268984.1 | 15 | 2046 | ||
GenBank mRNA | AK088950.1 | - | 14 | 3636 | ||
AK129220.1 | - | 14 | 3637 | |||
AK134073.1 | - | 3 | 1292 | |||
BC026492.1 | - | 8 | 991 | 4505095 | ||
BC092240.1 | - | 14 | 1957 | 30662573 |