Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tifab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168615.1 | NP_001162086.1 | 3 | 2032 | ||
NM_145976.4 | NP_666088.1 | 3 | 1968 | |||
GenBank mRNA | AK036643.1 | - | 3 | 1886 | ||
AK153654.1 | - | 3 | 1764 | |||
AK163959.1 | - | 1 | 998 | |||
BC005528.1 | - | 3 | 1676 | 3495346 | ||
BC027057.1 | - | 2 | 1551 | 5029696 | ||
BC031130.1 | - | 3 | 1800 | 4954852 |