Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163622.1 | NP_001157094.1 | 6 | 1114 | ||
NM_001163623.1 | NP_001157095.1 | 5 | 578 | |||
NM_001163624.1 | NP_001157096.1 | 5 | 446 | |||
NM_145984.3 | NP_666096.3 | 6 | 678 | |||
GenBank mRNA | AK028877.1 | - | 6 | 1072 | ||
AK050878.1 | - | 5 | 440 | |||
AK083070.1 | - | 6 | 619 | |||
AK163108.1 | - | 6 | 1079 | |||
BC004612.1 | - | 5 | 532 | 3585402 | ||
BC129880.1 | - | 4 | 1110 | |||
BY257815.1 | - | 5 | 447 |