Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146008.2 | NP_666120.1 | 5 | 754 | ||
GenBank mRNA | AK040850.1 | - | 5 | 805 | ||
AK088802.1 | - | 1 | 77 | |||
AK146035.1 | - | 5 | 1079 | |||
BC008239.1 | - | 4 | 551 | |||
BC029663.1 | - | 5 | 754 | 5364434 |