Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146039.3 | NP_666151.3 | 3 | 258 | ||
GenBank mRNA | AK032986.1 | - | 3 | 220 | ||
AK046002.1 | - | 4 | 358 | |||
AK046399.1 | - | 3 | 220 | |||
AK052482.1 | - | 3 | 222 | |||
AK079764.1 | - | 3 | 213 | |||
AK140242.1 | - | 4 | 358 | |||
AK144138.1 | - | 3 | 258 | |||
BC138916.1 | - | 3 | 191 | |||
CJ150229.1 | - | 3 | 258 |