Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146047.2 | NP_666159.2 | 2, 3 | 507 | ||
GenBank mRNA | AK032295.1 | - | 2, 3 | 470 | ||
AK048357.1 | - | 2, 3 | 404 | |||
AK156203.1 | - | 2, 3 | 398 | |||
AK165939.1 | - | 2, 3 | 367 | |||
AK178755.1 | - | 1, 2 | 44 | |||
AK192551.1 | - | 22 | ||||
AK220105.1 | - | 1, 2 | 44 | |||
BC026562.1 | - | 2, 3 | 208 | 5028264 |