Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146094.2 | NP_666206.1 | 1 | 304 | ||
GenBank mRNA | AB072976.1 | - | 1 | 219 | ||
AF061504.1 | - | 1 | 206 | |||
AK080706.1 | - | 1 | 208 | |||
AK083959.1 | - | 1 | 166 | |||
AK154367.1 | - | 1 | 254 | |||
BC022139.1 | - | 1 | 249 | 5100199 | ||
BC026831.1 | - | 1 | 271 | 4239695 | ||
BC026848.1 | - | 1 | 244 | 4196030 | ||
BC063053.1 | - | 1 | 164 | 5696722 | ||
KF987079.1 | - | 1 | 188 |